siRNA Details
| Gene Symbol: Rasgrp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006239757.2 | XP_006239819.1 | 13, 14 | 1713 | ||
| XM_006239758.2 | XP_006239820.1 | 14, 15 | 2150 | |||
| XM_006239759.2 | XP_006239821.1 | 13, 14 | 2042 | |||
| XM_006239760.2 | XP_006239822.1 | 13, 14 | 2017 | |||
| XM_006239761.2 | XP_006239823.1 | 11, 12 | 1538 | |||
| XM_006239762.2 | XP_006239824.1 | 12, 13 | 1909 | |||
| XM_008764485.1 | XP_008762707.1 | 9, 10 | 2208 | |||

Rasgrp3
Genomic Map