siRNA Details
| Gene Symbol: Mboat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108016.2 | NP_001101486.2 | 12 | 1399 | ||
| XM_006239985.2 | XP_006240047.1 | 11 | 1273 | |||
| GenBank mRNA | DV718174.1 | - | 2 | 221 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mboat2| Gene Symbol: Mboat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108016.2 | NP_001101486.2 | 12 | 1399 | ||
| XM_006239985.2 | XP_006240047.1 | 11 | 1273 | |||
| GenBank mRNA | DV718174.1 | - | 2 | 221 | ||