This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CMPK2 SIRNA Assays ›
Gene Symbol:
Cmpk2
Gene Name:
cytidine monophosphate (UMP-CMP) kinase 2, mitochondrial
Gene Aliases:
Tyki
Chromosome Location:
Chr.6: 45683242 - 45694824 on Build Rnor_6.0
UniGene ID:
Rn.100440
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s162949

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cmpk2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108017.1 NP_001101487.1 1 627
XM_006239938.2 XP_006240000.1 2 1314
XM_006239939.2 XP_006240001.1 2 1314

Back To Top

More Information


Panther Classification:

Molecular Function -

nucleotide kinase

Gene Ontology Categories:

Function(s) Process(es)

nucleoside diphosphate phosphorylation
dUDP biosynthetic process
dTDP biosynthetic process
dTTP biosynthetic process
nucleoside triphosphate biosynthetic process
nucleotide phosphorylation
cellular response to lipopolysaccharide
cytidylate kinase activity
nucleoside diphosphate kinase activity
thymidylate kinase activity
uridylate kinase activity
UMP kinase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-rdsp8:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline