This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PCNX SIRNA Assays ›
Gene Symbol:
Pcnx
Gene Name:
pecanex homolog (Drosophila)
Gene Aliases:
-
Chromosome Location:
Chr.6: 105611430 - 105747176 on Build Rnor_6.0
UniGene ID:
Rn.23381
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s163090

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pcnx
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170347.1 NP_001163818.1 2 183
XM_006240299.2 XP_006240361.1 2 209
XM_006240300.2 XP_006240362.1 2 209
XM_006240301.1 XP_006240363.1 3 245
XM_006240302.2 XP_006240364.1 2 209
XM_008764788.1 XP_008763010.1 2 209
XM_008764789.1 XP_008763011.1 2 1605

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-dpz9l:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0