siRNA Details
| Gene Symbol: RGD1310852 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025007.1 | NP_001020178.1 | 6 | 1016 | ||
| GenBank mRNA | BC091232.1 | - | 6 | 1016 | 7322456 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1310852| Gene Symbol: RGD1310852 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025007.1 | NP_001020178.1 | 6 | 1016 | ||
| GenBank mRNA | BC091232.1 | - | 6 | 1016 | 7322456 | |