This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other LAPTM4B SIRNA Assays ›
Gene Symbol:
Laptm4b
Gene Name:
lysosomal protein transmembrane 4 beta
Gene Aliases:
-
Chromosome Location:
Chr.7: 72924911 - 72968099 on Build Rnor_6.0
UniGene ID:
Rn.8123
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s163639

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Laptm4b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013174.1 NP_001013192.1 1, 2 169
GenBank mRNA BC086442.1 - 1, 2 169 7303600

Back To Top

More Information


Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s)

transport

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-lwb2x:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0