siRNA Details
| Gene Symbol: Mfng | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199110.1 | NP_954541.1 | 1 | 189 | ||
| GenBank mRNA | BC061801.1 | - | 1 | 189 | 5598335 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mfng| Gene Symbol: Mfng | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199110.1 | NP_954541.1 | 1 | 189 | ||
| GenBank mRNA | BC061801.1 | - | 1 | 189 | 5598335 | |