This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FBLN1 SIRNA Assays ›
Gene Symbol:
Fbln1
Gene Name:
fibulin 1
Gene Aliases:
-
Chromosome Location:
Chr.7: 126096793 - 126176468 on Build Rnor_6.0
UniGene ID:
Rn.9375
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s163800

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fbln1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127547.1 NP_001121019.1 8, 9 1049
XM_006242161.2 XP_006242223.1 8, 9 1029
GenBank mRNA BC161973.1 - 8, 9 1011 9029287

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_010180, EU543224, X70854, AK137511, AK188438, and others.

Gene Ontology Categories:

Function(s) Process(es)

embryo implantation
positive regulation of peptidase activity
extracellular matrix organization
molecular_function
calcium ion binding
peptidase activator activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-ndfxr:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline