siRNA Details
| Gene Symbol: Adamts7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047101.1 | NP_001040566.1 | 10 | 1948 | ||
| XM_006243523.1 | XP_006243585.1 | 9 | 1606 | |||
| XM_006243524.1 | XP_006243586.1 | 9 | 1606 | |||
| XM_006243525.1 | XP_006243587.1 | 9 | 1606 | |||
| XM_006243526.1 | XP_006243588.1 | 9 | 1606 | |||
| XM_006243527.2 | XP_006243589.1 | 9 | 3873 | |||
| XM_008766447.1 | XP_008764669.1 | 9 | 1981 | |||
| GenBank mRNA | AY257482.1 | - | 10 | 1948 | ||
| AY327121.1 | - | 9 | 1366 | |||

Adamts7
Genomic Map