siRNA Details
| Gene Symbol: Ky | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108180.1 | NP_001101650.1 | 10 | 1388 | ||
| XM_006243657.2 | XP_006243719.1 | 10 | 1430 | |||
| XM_006243658.1 | XP_006243720.1 | 7 | 881 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ky| Gene Symbol: Ky | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108180.1 | NP_001101650.1 | 10 | 1388 | ||
| XM_006243657.2 | XP_006243719.1 | 10 | 1430 | |||
| XM_006243658.1 | XP_006243720.1 | 7 | 881 | |||