This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PIKFYVE SIRNA Assays ›
Gene Symbol:
Pikfyve
Gene Name:
phosphoinositide kinase, FYVE finger containing
Gene Aliases:
Pip5k3
Chromosome Location:
Chr.9: 71911748 - 72005816 on Build Rnor_6.0
UniGene ID:
Rn.205590
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s164874

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 400.00
Your Price
Online offer: 388.65
388.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pikfyve
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006245071.2 XP_006245133.1 6 937
XM_006245072.2 XP_006245134.1 6 934
XM_006245073.2 XP_006245135.1 5 901
XM_006245074.2 XP_006245136.1 6 797
XM_006245076.2 XP_006245138.1 6 742
XM_006245077.2 XP_006245139.1 7 772
XM_006245078.2 XP_006245140.1 6 937
XM_006245079.2 XP_006245141.1 6 934
XM_008767169.1 XP_008765391.1 6 3938
XM_008767170.1 XP_008765392.1 6 937

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011086, AK173077, AF102777, AK165350, AK080527, and others.

Panther Classification:

Molecular Function -

kinase

Gene Ontology Categories:

Function(s) Process(es)

receptor-mediated endocytosis
myelin assembly
protein localization to nucleus
intracellular signal transduction
retrograde transport, endosome to Golgi
cellular protein metabolic process
phosphatidylinositol phosphorylation
ATP binding
zinc ion binding
phosphatidylinositol phosphate kinase activity
1-phosphatidylinositol-4-phosphate 5-kinase activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-4dqkp:80/100.66.77.166:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0