siRNA Details
| Gene Symbol: Trip12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031659.1 | NP_001026829.1 | 15 | 2314 | ||
| XM_006245377.2 | XP_006245439.1 | 16 | 2420 | |||
| XM_006245379.2 | XP_006245441.1 | 16 | 2420 | |||
| XM_006245380.2 | XP_006245442.1 | 16 | 2420 | |||
| XM_006245386.1 | XP_006245448.1 | 15 | 2339 | |||
| XM_008767252.1 | XP_008765474.1 | 16 | 2420 | |||
| XM_008767253.1 | XP_008765475.1 | 16 | 2471 | |||
| XM_008767254.1 | XP_008765476.1 | 16 | 2420 | |||
| XM_008767255.1 | XP_008765477.1 | 16 | 2420 | |||
| XM_008767256.1 | XP_008765478.1 | 17 | 2333 | |||
| XM_008767257.1 | XP_008765479.1 | 15 | 2339 | |||
| GenBank mRNA | BC100624.1 | - | 15 | 2314 | 7930421 | |

Trip12
Genomic Map