siRNA Details
| Gene Symbol: Magix | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014109.1 | NP_001014131.1 | 2, 3 | 365 | ||
| GenBank mRNA | BC078960.1 | - | 2, 3 | 365 | 7110859 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Magix| Gene Symbol: Magix | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014109.1 | NP_001014131.1 | 2, 3 | 365 | ||
| GenBank mRNA | BC078960.1 | - | 2, 3 | 365 | 7110859 | |