This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other USP51 SIRNA Assays ›
Gene Symbol:
Usp51
Gene Name:
ubiquitin specific peptidase 51
Gene Aliases:
RGD1562211
Chromosome Location:
Chr.X: 19268499 - 19273753 on Build Rnor_6.0
UniGene ID:
Rn.56303
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s165275

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Usp51
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108252.1 NP_001101722.1 2 1871
XM_006256768.2 XP_006256830.1 4 2203
XM_008773076.1 XP_008771298.1 3 2941

Back To Top

More Information


Panther Classification:

Molecular Function -

cysteine protease

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
histone deubiquitination
protein deubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
regulation of proteasomal protein catabolic process
cysteine-type endopeptidase activity
thiol-dependent ubiquitin-specific protease activity
zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0