This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GTF2I SIRNA Assays ›
Gene Symbol:
Gtf2i
Gene Name:
general transcription factor II I
Gene Aliases:
Gtf2ird1
Chromosome Location:
Chr.12: 25410804 - 25487970 on Build Rnor_6.0
UniGene ID:
Rn.27575
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s165532

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gtf2i
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001512.2 NP_001001512.2 13 1291
XM_006249162.2 XP_006249224.1 17 1556
XM_006249163.2 XP_006249225.1 17 1553
XM_006249164.2 XP_006249226.1 16 1496
XM_006249165.2 XP_006249227.1 16 1493
XM_006249166.2 XP_006249228.1 16 1490
XM_006249167.2 XP_006249229.1 15 1439
XM_006249168.2 XP_006249230.1 15 1433
XM_006249169.2 XP_006249231.1 15 1418
XM_006249170.2 XP_006249232.1 17 1751
XM_006249171.2 XP_006249233.1 14 1376
XM_006249172.2 XP_006249234.1 14 1361
XM_006249173.2 XP_006249235.1 14 1355
XM_006249174.2 XP_006249236.1 13 1298
XM_008769141.1 XP_008767363.1 17 1418
GenBank mRNA AF547390.1 - 15 1328
AF547391.1 - 15 1328
BC085815.1 - 13 1429 7123295

Back To Top

More Information


Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
transition between slow and fast fiber
negative regulation of angiogenesis
negative regulation of cytosolic calcium ion concentration
molecular_function
DNA binding
mitogen-activated protein kinase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-f5rhb:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0