This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GTF2I SIRNA Assays ›
Gene Symbol:
Gtf2i
Gene Name:
general transcription factor II I
Gene Aliases:
Gtf2ird1
Chromosome Location:
Chr.12: 25410804 - 25487970 on Build Rnor_6.0
UniGene ID:
Rn.27575
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s165533

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gtf2i
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001512.2 NP_001001512.2 24 2437
XM_006249162.2 XP_006249224.1 28 2702
XM_006249163.2 XP_006249225.1 28 2699
XM_006249164.2 XP_006249226.1 27 2642
XM_006249165.2 XP_006249227.1 27 2639
XM_006249166.2 XP_006249228.1 27 2636
XM_006249167.2 XP_006249229.1 26 2585
XM_006249168.2 XP_006249230.1 26 2579
XM_006249169.2 XP_006249231.1 26 2564
XM_006249170.2 XP_006249232.1 28 2897
XM_006249171.2 XP_006249233.1 25 2522
XM_006249172.2 XP_006249234.1 25 2507
XM_006249173.2 XP_006249235.1 25 2501
XM_006249174.2 XP_006249236.1 24 2444
XM_008769141.1 XP_008767363.1 28 2564
GenBank mRNA AF547390.1 - 26 2474
AF547391.1 - 26 2474
BC085815.1 - 24 2575 7123295

Back To Top

More Information


Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
transition between slow and fast fiber
negative regulation of angiogenesis
negative regulation of cytosolic calcium ion concentration
molecular_function
DNA binding
mitogen-activated protein kinase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0