siRNA Details
| Gene Symbol: Ksr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108284.1 | NP_001101754.1 | 6, 7 | 1162 | ||
| XM_006246924.2 | XP_006246986.1 | 7, 8 | 1385 | |||
| XM_006246926.2 | XP_006246988.1 | 7, 8 | 1383 | |||
| XM_006246927.2 | XP_006246989.1 | 7, 8 | 1383 | |||
| XM_008768063.1 | XP_008766285.1 | 7, 8 | 1386 | |||
| XM_008768064.1 | XP_008766286.1 | 7, 8 | 1385 | |||
| XM_008768065.1 | XP_008766287.1 | 7, 8 | 1385 | |||
| XM_008768066.1 | XP_008766288.1 | 6, 7 | 1361 | |||
| XM_008768067.1 | XP_008766289.1 | 7, 8 | 1384 | |||

Ksr1
Genomic Map