This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CDC27 SIRNA Assays ›
Gene Symbol:
Cdc27
Gene Name:
cell division cycle 27
Gene Aliases:
-
Chromosome Location:
Chr.10: 92551754 - 92602217 on Build Rnor_6.0
UniGene ID:
Rn.198335
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s166011

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cdc27
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024793.1 NP_001019964.1 15 2007
XM_006247533.2 XP_006247595.1 14 3728
XM_006247534.2 XP_006247596.1 14 3725
XM_006247535.2 XP_006247597.1 15 2144
XM_006247536.2 XP_006247598.1 13 3576
XM_006247537.1 XP_006247599.1 12 1676
XM_008768295.1 XP_008766517.1 14 1997
GenBank mRNA BC097475.1 - 15 2007 7457819

Back To Top

More Information


Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

metaphase/anaphase transition of mitotic cell cycle
protein K11-linked ubiquitination
protein phosphatase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0