This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TIPRL SIRNA Assays ›
Gene Symbol:
Tiprl
Gene Name:
TOR signaling pathway regulator
Gene Aliases:
RGD1310442
Chromosome Location:
Chr.13: 83478966 - 83504448 on Build Rnor_6.0
UniGene ID:
Rn.56867
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s166447

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 400.00
Your Price
Online offer: 296.00
296.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tiprl
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109667.1 NP_001103137.1 4 453
GenBank mRNA BC128780.1 - 4 453 7315719

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_001031800, NM_152902, AL049670, BC009506, AK313520, and others.
This siRNA also targets the following transcripts in Mouse: NM_145513, AK049119, BC058250, BC002098, AK049784, and others.

Gene Ontology Categories:

Function(s)

DNA damage checkpoint
negative regulation of protein phosphatase type 2A activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-5mz9h:80/100.66.72.156:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0