siRNA Details
| Gene Symbol: Tmed9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009703.1 | NP_001009703.1 | 3, 4 | 426 | ||
| GenBank mRNA | BC088422.1 | - | 3, 4 | 426 | 7123664 | |
| FQ219234.1 | - | 3, 4 | 434 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmed9| Gene Symbol: Tmed9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009703.1 | NP_001009703.1 | 3, 4 | 426 | ||
| GenBank mRNA | BC088422.1 | - | 3, 4 | 426 | 7123664 | |
| FQ219234.1 | - | 3, 4 | 434 | |||