This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TOM1 SIRNA Assays ›
Gene Symbol:
Tom1
Gene Name:
target of myb1 (chicken)
Gene Aliases:
-
Chromosome Location:
Chr.19: 14392367 - 14426482 on Build Rnor_6.0
UniGene ID:
Rn.198076
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s167237

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tom1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008365.1 NP_001008366.1 3 193
XM_006255145.2 XP_006255207.1 3 303
GenBank mRNA BC083873.1 - 3 193 7128979

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001136259, NM_011622, AK160403, AK028398, AK150143, and others.

Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
clathrin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-x666z:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline