This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other EDC4 SIRNA Assays ›
Gene Symbol:
Edc4
Gene Name:
enhancer of mRNA decapping 4
Gene Aliases:
LRRG00115, Nrn1l, RGD1309136
Chromosome Location:
Chr.19: 37852659 - 37864651 on Build Rnor_6.0
UniGene ID:
Rn.145173
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s167283

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Edc4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033068.3 NP_001028240.3 23 3325
GenBank mRNA BC103628.1 - 23 3325 7939290

Back To Top

More Information


Panther Classification:

Molecular Function -

mRNA capping factor

Gene Ontology Categories:

Function(s)

nervous system development

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0