This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FANCA SIRNA Assays ›
Gene Symbol:
Fanca
Gene Name:
Fanconi anemia, complementation group A
Gene Aliases:
-
Chromosome Location:
Chr.19: 56067548 - 56126074 on Build Rnor_6.0
UniGene ID:
Rn.163415
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s167355

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fanca
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108455.1 NP_001101925.1 21, 22 1924
XM_008772633.1 XP_008770855.1 22, 23 2048

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

male meiosis
biological_process
male gonad development
female gonad development
regulation of cell proliferation
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-gm74s:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline