siRNA Details
| Gene Symbol: Me3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108491.2 | NP_001101961.2 | 15 | 1795 | ||
| XM_006229666.1 | XP_006229728.1 | 14 | 1693 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Me3| Gene Symbol: Me3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108491.2 | NP_001101961.2 | 15 | 1795 | ||
| XM_006229666.1 | XP_006229728.1 | 14 | 1693 | |||