siRNA Details
| Gene Symbol: Pitpnm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008369.1 | NP_001008370.1 | 8 | 1313 | ||
| GenBank mRNA | BC085945.1 | - | 8 | 1313 | 7128360 | |
| Gene Symbol: Pde4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | M26717.1 | - | 71 | |||

Pitpnm1
Genomic Map