siRNA Details
| Gene Symbol: Smarca2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004446.1 | NP_001004446.1 | 18 | 2714 | ||
| XM_006231227.2 | XP_006231289.1 | 18 | 2732 | |||
| XM_008760331.1 | XP_008758553.1 | 18 | 2992 | |||
| XM_008760332.1 | XP_008758554.1 | 19 | 3040 | |||
| XM_008760333.1 | XP_008758555.1 | 19 | 2780 | |||
| XM_008760334.1 | XP_008758556.1 | 18 | 2812 | |||
| XM_008760335.1 | XP_008758557.1 | 18 | 2748 | |||
| XM_008760336.1 | XP_008758558.1 | 18 | 3060 | |||
| XM_008760337.1 | XP_008758559.1 | 17 | 2610 | |||
| XM_008760338.1 | XP_008758560.1 | 16 | 2246 | |||
| GenBank mRNA | AY643746.1 | - | 18 | 2714 | ||

Smarca2
Genomic Map