This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other HNRNPA3 SIRNA Assays ›
Gene Symbol:
Hnrnpa3
Gene Name:
heterogeneous nuclear ribonucleoprotein A3
Gene Aliases:
Hnrpa3
Chromosome Location:
Chr.3: 62481228 - 62491356 on Build Rnor_6.0
UniGene ID:
Rn.107690
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s168604

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Hnrnpa3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001111294.1 NP_001104764.1 9 1105
NM_001111295.1 NP_001104765.1 9 1039
NM_198132.3 NP_937765.1 9 1105
XM_008761911.1 XP_008760133.1 9 1054
XM_008761912.1 XP_008760134.1 9 1054
GenBank mRNA AY363226.1 - 9 1051
AY363227.1 - 9 985
BC081878.1 - 9 1045 7108128
CF110891.1 - 3 271
FQ214617.1 - 9 1001
FQ220486.1 - 2 89
Y16641.1 - 5 467

Back To Top

More Information


Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
RNA splicing
mRNA transport
nucleotide binding
nucleic acid binding
mRNA binding
poly(A) RNA binding
RNA transmembrane transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-jrfm9:80/100.66.76.154:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0