This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CWC22 SIRNA Assays ›
Gene Symbol:
Cwc22
Gene Name:
CWC22 spliceosome-associated protein
Gene Aliases:
RGD1565385
Chromosome Location:
Chr.3: 64731145 - 64780127 on Build Rnor_6.0
UniGene ID:
Rn.51454
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s168606

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 400.00
Your Price
Online offer: 296.00
296.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cwc22
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006234425.2 XP_006234487.1 7 934
XM_008761965.1 XP_008760187.1 7 937
XM_008761966.1 XP_008760188.1 7 924
XM_008761967.1 XP_008760189.1 7 934
XM_342453.7 XP_342454.3 7 936
GenBank mRNA FQ231274.1 - 7 864
FQ231579.1 - 7 864

Back To Top

More Information


Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Process(es)

RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mjsjv:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline