This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TMUB1 SIRNA Assays ›
Gene Symbol:
Tmub1
Gene Name:
transmembrane and ubiquitin-like domain containing 1
Gene Aliases:
Hops
Chromosome Location:
Chr.4: 7257016 - 7260318 on Build Rnor_6.0
UniGene ID:
Rn.105624
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s168893

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tmub1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080153.1 NP_001073622.1 2 304
NM_198781.2 NP_942076.2 2 214
XM_006235944.2 XP_006236006.1 2 950
XM_006235945.2 XP_006236007.1 2 341
XM_008762651.1 XP_008760873.1 2 216
GenBank mRNA AY603378.1 - 2 304
DY313123.1 - 2 214

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0