siRNA Details
| Gene Symbol: Creb3l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012188.1 | NP_001012188.1 | 4 | 888 | ||
| GenBank mRNA | AY546001.1 | - | 4 | 888 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Creb3l2| Gene Symbol: Creb3l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012188.1 | NP_001012188.1 | 4 | 888 | ||
| GenBank mRNA | AY546001.1 | - | 4 | 888 | ||