This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other UBAP1 SIRNA Assays ›
Gene Symbol:
Ubap1
Gene Name:
ubiquitin-associated protein 1
Gene Aliases:
UBAP-1
Chromosome Location:
Chr.5: 57738312 - 57778724 on Build Rnor_6.0
UniGene ID:
Rn.98217
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s169225

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ubap1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012190.1 NP_001012190.1 4 708
XM_008763626.1 XP_008761848.1 4 717
GenBank mRNA BC083594.1 - 4 708 7132783

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein transport
ubiquitin-dependent protein catabolic process via the multivesicular body sorting pathway
ubiquitin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-ndfxr:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline