siRNA Details
| Gene Symbol: Kpna6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015029.2 | NP_001015029.2 | 12 | 1341 | ||
| GenBank mRNA | AY779030.1 | - | 12 | 1291 | ||
| BC105813.1 | - | 12 | 1341 | 7112648 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kpna6| Gene Symbol: Kpna6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015029.2 | NP_001015029.2 | 12 | 1341 | ||
| GenBank mRNA | AY779030.1 | - | 12 | 1291 | ||
| BC105813.1 | - | 12 | 1341 | 7112648 | ||