siRNA Details
| Gene Symbol: Brd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100903.1 | NP_001094373.1 | 10 | 2082 | ||
| XM_006241072.2 | XP_006241134.1 | 10 | 2086 | |||
| XM_006241073.2 | XP_006241135.1 | 10 | 2290 | |||
| XM_006241074.2 | XP_006241136.1 | 10 | 3014 | |||
| XM_008765170.1 | XP_008763392.1 | 9 | 3805 | |||
| XM_008765171.1 | XP_008763393.1 | 9 | 3805 | |||
| XM_008765172.1 | XP_008763394.1 | 9 | 3805 | |||
| XM_008765173.1 | XP_008763395.1 | 8 | 2332 | |||

Brd4
Genomic Map