siRNA Details
| Gene Symbol: Adck5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135798.1 | NP_001129270.1 | 10 | 1132 | ||
| XM_006241827.2 | XP_006241889.1 | 10 | 1036 | |||
| GenBank mRNA | BC168693.1 | - | 10 | 1120 | 7935927 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adck5| Gene Symbol: Adck5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135798.1 | NP_001129270.1 | 10 | 1132 | ||
| XM_006241827.2 | XP_006241889.1 | 10 | 1036 | |||
| GenBank mRNA | BC168693.1 | - | 10 | 1120 | 7935927 | |