siRNA Details
| Gene Symbol: RGD1306001 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130696.1 | NP_001124168.1 | 4 | 357 | ||
| XM_006242163.1 | XP_006242225.1 | 4 | 933 | |||
| XM_008765765.1 | XP_008763987.1 | 4 | 521 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1306001| Gene Symbol: RGD1306001 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130696.1 | NP_001124168.1 | 4 | 357 | ||
| XM_006242163.1 | XP_006242225.1 | 4 | 933 | |||
| XM_008765765.1 | XP_008763987.1 | 4 | 521 | |||