siRNA Details
| Gene Symbol: Rnase1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013232.1 | NP_001013250.1 | 1 | 113 | ||
| XM_006251893.2 | XP_006251955.1 | 2 | 244 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnase1l1| Gene Symbol: Rnase1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013232.1 | NP_001013250.1 | 1 | 113 | ||
| XM_006251893.2 | XP_006251955.1 | 2 | 244 | |||