siRNA Details
| Gene Symbol: RGD1311300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF196199.1 | - | 2 | 332 | ||
| BC099216.1 | - | 2 | 340 | 7384680 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1311300| Gene Symbol: RGD1311300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF196199.1 | - | 2 | 332 | ||
| BC099216.1 | - | 2 | 340 | 7384680 | ||