siRNA Details
| Gene Symbol: Blk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025751.1 | NP_001020922.1 | 10 | 1317 | ||
| XM_006252188.2 | XP_006252250.1 | 11 | 1711 | |||
| XM_008770769.1 | XP_008768991.1 | 10 | 1738 | |||
| XM_008770770.1 | XP_008768992.1 | 9 | 977 | |||
| GenBank mRNA | BC098683.1 | - | 10 | 1317 | 7385175 | |

Blk
Genomic Map