This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SYCE2 SIRNA Assays ›
Gene Symbol:
Syce2
Gene Name:
synaptonemal complex central element protein 2
Gene Aliases:
RGD1311867
Chromosome Location:
Chr.19: 25981601 - 26002399 on Build Rnor_6.0
UniGene ID:
Rn.230993
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s171776

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Syce2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191557.1 NP_001178486.1 2 228
XM_006255291.2 XP_006255353.1 3 1778

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_027954, NM_001168246, NM_001168244, AK167332, BC094628, and others.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-2x92s:80/100.66.79.168:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0