siRNA Details
| Gene Symbol: Scoc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013235.1 | NP_001013253.1 | 1 | 65 | ||
| XM_006255292.2 | XP_006255354.1 | 1 | 216 | |||
| GenBank mRNA | BC076390.1 | - | 1 | 65 | 7106066 | |
| BC086417.1 | - | 1 | 65 | 7127948 | ||

Scoc
Genomic Map