siRNA Details
| Gene Symbol: Zfp524 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108905.1 | NP_001102375.1 | 1 | 168 | ||
| XM_008758958.1 | XP_008757180.1 | 2 | 266 | |||
| XM_008758959.1 | XP_008757181.1 | 2 | 194 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp524| Gene Symbol: Zfp524 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108905.1 | NP_001102375.1 | 1 | 168 | ||
| XM_008758958.1 | XP_008757180.1 | 2 | 266 | |||
| XM_008758959.1 | XP_008757181.1 | 2 | 194 | |||