This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZFP524 SIRNA Assays ›
Gene Symbol:
Zfp524
Gene Name:
zinc finger protein 524
Gene Aliases:
RGD1565485, Znf524
Chromosome Location:
Chr.1: 72376381 - 72379883 on Build Rnor_6.0
UniGene ID:
Rn.24795
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s171884

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zfp524
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108905.1 NP_001102375.1 1 298
XM_008758958.1 XP_008757180.1 2 396
XM_008758959.1 XP_008757181.1 2 324

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_025324, AK009053, BC019995, AK002594, AK011620, and others.

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-tc7l7:80/100.66.79.14:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0