siRNA Details
| Gene Symbol: Trim55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012218.1 | NP_001012218.1 | 7 | 1068 | ||
| XM_006232181.2 | XP_006232243.1 | 7 | 1106 | |||
| XM_006232182.2 | XP_006232244.1 | 7 | 1106 | |||
| GenBank mRNA | BC087100.1 | - | 7 | 1068 | 7192411 | |

Trim55
Genomic Map