This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other LOC365778 SIRNA Assays ›
Gene Symbol:
LOC365778
Gene Name:
similar to RIKEN cDNA 1700034I23
Gene Aliases:
HCBP6
Chromosome Location:
Chr.2: 127776103 - 127778350 on Build Rnor_6.0
UniGene ID:
Rn.202583
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s172315

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LOC365778
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014251.1 NP_001014273.1 1 197
GenBank mRNA AY237129.1 - 49
BC078989.1 - 1 197 7112531

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: AK077188, AK006596, BC117739, AK077003, BY081654.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-5p4xw:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline