siRNA Details
| Gene Symbol: St6galnac5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271361.1 | NP_001258290.1 | 680 | |||
| GenBank mRNA | AJ646872.1 | - | 450 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

St6galnac5| Gene Symbol: St6galnac5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271361.1 | NP_001258290.1 | 680 | |||
| GenBank mRNA | AJ646872.1 | - | 450 | |||