This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DUS4L SIRNA Assays ›
Gene Symbol:
Dus4l
Gene Name:
dihydrouridine synthase 4-like (S. cerevisiae)
Gene Aliases:
RGD1311445
Chromosome Location:
Chr.6: 50929156 - 50943488 on Build Rnor_6.0
UniGene ID:
Rn.41975
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s172838

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dus4l
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135803.1 NP_001129275.1 3 310
XM_006239990.2 XP_006240052.1 3 241
XM_006239991.1 XP_006240053.1 4 468
XM_006239992.2 XP_006240054.1 4 745
XM_006239993.2 XP_006240055.1 2 154
XM_008764601.1 XP_008762823.1 2 116

Back To Top

More Information


Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Function(s) Process(es)

tRNA dihydrouridine synthesis
oxidation-reduction process
tRNA dihydrouridine synthase activity
flavin adenine dinucleotide binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gjvvj:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0