siRNA Details
| Gene Symbol: Tceal8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014275.1 | NP_001014297.1 | 3 | 353 | ||
| GenBank mRNA | AB108667.1 | - | 3 | 379 | ||
| BC086418.1 | - | 3 | 353 | 7128221 | ||
| FQ214154.1 | - | 3 | 449 | |||
| FQ214671.1 | - | 3 | 380 | |||
| FQ215624.1 | - | 3 | 407 | |||
| FQ224344.1 | - | 3 | 371 | |||
| FQ226647.1 | - | 3 | 376 | |||
| FQ233322.1 | - | 3 | 416 | |||
| FQ234425.1 | - | 3 | 420 | |||

Tceal8
Genomic Map