siRNA Details
| Gene Symbol: Olr1551 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000727.1 | NP_001000727.1 | 1 | 436 | ||
| XM_006248198.2 | XP_006248260.1 | 2 | 484 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1551| Gene Symbol: Olr1551 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000727.1 | NP_001000727.1 | 1 | 436 | ||
| XM_006248198.2 | XP_006248260.1 | 2 | 484 | |||