This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CCNB2 SIRNA Assays ›
Gene Symbol:
CCNB2
Gene Name:
cyclin B2
Gene Aliases:
HsT17299
Chromosome Location:
Chr.15: 59105085 - 59125045 on Build GRCh38
UniGene ID:
Hs.194698
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s17446
Validated

Purification
Size
Availability Inventoried
Catalog # 4390824
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CCNB2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004701.3 NP_004692.1 2 284
GenBank mRNA AB020981.1 - 2 169
AF002822.1 - 2 119
AF087910.1 - 2 226
AK001404.1 - 2 248
AK222611.1 - 2 217
AL080146.1 - 2 237
AM851054.1 - 2 100
BC105086.1 - 2 232 8144089
BC105112.1 - 2 232 8144115
BT019458.1 - 2 100
BU934242.1 - 2 285
CR533527.1 - 2 100

Back To Top

More Information


Matching Taqman Gene Expression Assay:


Hs00270424_m1

Panther Classification:

Molecular Function -

kinase activator

Gene Ontology Categories:

Function(s) Process(es)

G2/M transition of mitotic cell cycle
in utero embryonic development
mitotic nuclear division
mitotic nuclear envelope disassembly
growth
T cell homeostasis
thymus development
cell division
regulation of cell cycle
cell-cell adhesion
cyclin-dependent protein serine/threonine kinase activity
protein binding
cadherin binding involved in cell-cell adhesion

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mlg7h:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline