This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CCNB2 SIRNA Assays ›
Gene Symbol:
CCNB2
Gene Name:
cyclin B2
Gene Aliases:
HsT17299
Chromosome Location:
Chr.15: 59105085 - 59125045 on Build GRCh38
UniGene ID:
Hs.194698
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s17447
Validated

Purification
Size
Availability Inventoried
Catalog # 4390824
Price 556.00
Your Price
Online offer: 411.44
411.44
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CCNB2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004701.3 NP_004692.1 4 543
GenBank mRNA AB020981.1 - 4 428
AF002822.1 - 4 378
AF087910.1 - 4 485
AK001404.1 - 4 507
AK222611.1 - 4 476
AL080146.1 - 4 496
AM851054.1 - 4 359
BC105086.1 - 4 491 8144089
BC105112.1 - 4 491 8144115
BT019458.1 - 4 359
CR533527.1 - 4 359

Back To Top

More Information


Matching Taqman Gene Expression Assay:


Hs00270424_m1

Panther Classification:

Molecular Function -

kinase activator

Gene Ontology Categories:

Function(s) Process(es)

G2/M transition of mitotic cell cycle
in utero embryonic development
mitotic nuclear division
mitotic nuclear envelope disassembly
growth
T cell homeostasis
thymus development
cell division
regulation of cell cycle
cell-cell adhesion
cyclin-dependent protein serine/threonine kinase activity
protein binding
cadherin binding involved in cell-cell adhesion

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-cj8gb:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline